Imprimer
Avantor® Hichrom PEEKsil™, Chromatography Tubing, Stainless Steel
Fournisseur: Avantor
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
HICHHI-1109EA 126 EUR
HICHHI-1109 HICHHI-1128
Avantor® Hichrom PEEKsil™, Chromatography Tubing, Stainless Steel
Tuyaux Tubulures à chromatographie
Tubings are made of 316 stainless steel.
  • Resistant to chemicals
The precision bore is 1/16" type tubing.