Imprimer
Avantor® Hichrom, Fittings, Unions
Fournisseur: Avantor
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
HICHHI-055EA 63.4 EUR
HICHHI-055 HICHHI-059
Avantor® Hichrom, Fittings, Unions
Raccords et connecteurs
Fitings that are chemically inert and biocompatible
  • Zero dead volume connection
  • Pressure rated to 344 bar
  • Complete with two HI-505 fingertight fitting
Swept volume: 0,07 µl and 0,28 µl.