Imprimer
Pharmaceutical secondary standard, Stearyl Alcohol, certified reference material, Supelco®
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PHR1115-1GEA 84.8 EUR
PHR1115-1G
Pharmaceutical secondary standard, Stearyl Alcohol, certified reference material, Supelco®
Étalons Étalons organiques
Traceable to Ph. Eur. S1350000 and to USP 1622000.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Certifications: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.