Imprimer
VWR®, External PT100 Probes for
Circulating Baths
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
461-1373EA 490 EUR
461-1373 461-1374 461-1376 461-1377
VWR®, External PT100 Probes for
Circulating Baths
Sondes de température
External PT100 probes is for monitoring the temperature of an external application.
For use with: Circulating baths, refrigerated circulating baths, heating circulating baths, stainless steel open bath systems, polycarbonate open bath systems, specialty products, calibration bath, 75 L refrigerated circulating bath.