Imprimer
VWR®, Stand-up Sample Bags with Closure
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
129-1473EA 292 EUR
129-1473 129-1472
VWR®, Stand-up Sample Bags with Closure
Bags Sample Bags
These stand-up bags are meant for jobs where two hands are required.
  • Gamma sterilised
  • Maximum bag mouth opening to facilitate insertion of sample
  • Tear-off top which ensures internal sterility, eliminating the risk of internal contamination
The gusseted bags stand up by themselves without the necessity of a rack or stand. The combination of the flat bottom and the wire closure offers a bag that stands by itself and remains open until the sample has been poured in.
Informations de livraison: Sterility confirmation documentation available in every box.