Imprimer
Avantor® Hichrom Bottom-of-the-Bottle™, Inlet Solvent Filters
Fournisseur: Avantor
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
HICHHI-579EA 52.9 EUR
HICHHI-579 HICHHI-580
Avantor® Hichrom Bottom-of-the-Bottle™, Inlet Solvent Filters
Filters Liquid Process Filters
These filters are made from 100% PEEK.
  • Biocompatible filters
  • Two built-in PEEK frits
The bottom frit (2 or 10 µm) will draw solvents from within 2 mm of the bottom of the solvent bottle. The 2 μm frit on the side may be used for a Ext.Ø 203 mm helium sparging line.