Imprimer
Magnetic spin vanes for use with NextGen™ V Vials®, WHEATON®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
903061 903063
WHEA903061EA 114 EUR
WHEA903061 WHEA903063
Magnetic spin vanes for use with NextGen™ V Vials®, WHEATON®
Agitateurs Barreaux magnétiques
These stirring bars are 'V' shaped and are constructed with a cross-mounted, moulded-in magnetic bar.
The stir bars are PTFE-coated for use with aggressive chemical samples and are heat and chemically resistant.