Imprimer
Avantor® Partisil ODS3, HPLC Columns
Fournisseur: Avantor
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
P10ODS3-250AEA 1150 EUR
P10ODS3-250A P5ODS3-150A P5ODS3-250A P10ODS3-10C5 P5ODS3-10C5 P10ODS3-150A P5ODS3-100A P5ODS3-250AF P5ODS3-125A P10ODS3-300AF
Avantor® Partisil ODS3, HPLC Columns
Colonnes pour chromatographie
Avantor® Partisil® was one of the first commercially available irregular silicas with a large surface area giving it a high loading capacity.
  • Easy to use
  • Irregular porous silica
Avantor® Partisil® ODS3 is an intermediate carbon load C18 phase used for pharmaceutical, natural products, food, biological and environmental pollutants applications.