Imprimer
Avantor® Alltima HP C18, HPLC Columns, 5 µm
Fournisseur: Avantor
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
HICH87665EA 893 EUR
HICH87665 HICH87681 HICH87682 HICH87602 HICH87603 HICH87643 HICH87679 HICH87680 87683/N 87684/N
Avantor® Alltima HP C18, HPLC Columns, 5 µm
Colonnes pour chromatographie
Avantor® Alltima HP phase C18 columns are classic reverse phase retention and selectivity.
  • Better stability
  • High purity
  • Excellent column performance
  • pH: 1 to 10
Multiple selectivity options for optimising retention, resolution and analysis time. High purity silica helps to eliminate peak tailing problems.
Low column bleed makes the column to be ideal for microbore and other critical analysis.