Imprimer
Sigma Pseudo™ Corpse Scent, Formulation II, Supelco®
Marques : Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
P3929-20ML P3929-8X1ML
P3929-20MLEA 253 EUR
P3929-20ML P3929-8X1ML
Sigma Pseudo™ Corpse Scent, Formulation II, Supelco®
Étalons Étalons organiques
Canine training aids for the detection of corpses. For post-putrification detection.
Sigma pseudo™ corpse scent may be used as an effective artificial scent in training cadaver dogs. The volatile compounds of its decomposition have been analyzed using gas chromatography coupled with time of flight mass spectrometry (GC–TOFMS).