Imprimer
Density standards, H&D Fitzgerald Ltd. Quality, certified reference materials, Supelco®
Marques : Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
44964-10ML-F 12156-10ML-F 36232-10ML 76731-10ML
44964-10ML-FEA 237 EUR
44964-10ML-F 12156-10ML-F 36232-10ML 76731-10ML
Density standards, H&D Fitzgerald Ltd. Quality, certified reference materials, Supelco®
Étalons Étalons de densité
The exact density values can be taken from the certificate of analysis.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d42fb71b4903903105c9fce7fd0b6e70892d5f44&vid=d42fb71b4903903105c9fce7fd0b6e70892d5f44&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=354fb24d51e5fda6a269f81e783dc3e28deb3541&vid=354fb24d51e5fda6a269f81e783dc3e28deb3541&prid=web&clid=SAPDAM