Imprimer
Round bottom flasks with standard glass ground joint
Fournisseur: witeg Labortechnik
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0.618.002 0.618.500 0.615.025 0.615.050 0.618.001 0.618.050 0.618.100 0.618.250
201-1368EA 143 EUR
201-1368 201-1299 201-1293 201-1294 201-1295 201-1296 201-1297 201-1298
Round bottom flasks with standard glass ground joint
Ballons Flacons à fond rond
DURAN® borosilicate glass 3.3, amber.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With centre neck NS 14/23 or NS 29/32 and coloured brown to protect light-sensitive substances.
Certifications: According to DIN 12348 specifications.