Imprimer
VWR®, TLC Glass Plates, Silica HP, Unmodified Layers
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
551-0007EA 191 EUR
551-0007 551-0012 551-0015 551-0016 551-0025
VWR®, TLC Glass Plates, Silica HP, Unmodified Layers
TLC Plates
These TLC glass plates have an excellent separation efficiency due to an optimised particle size distribution.
  • Specific dyeability and abrasion resistance based on an optimised binder system
  • Good wettability for exact colorisation results, even with 100% aqueous detection reagents
Specific surface (BET): ~500 m²/g
Specific pore volume: 0,75 ml/g
Particle size: 5 - 17 µm