Imprimer
Organic reference standard, Valerenic acid, analytical standard, Supelco®
Marques : Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
51964-1MG 51964-5MG
51964-1MGEA 82.2 EUR
51964-1MG 51964-5MG
Organic reference standard, Valerenic acid, analytical standard, Supelco®
Étalons Étalons organiques
Valerenic acid is a bioactive sesquiterpene and a natural product, isolated from Valeriana officinalis type of plant.
Cdt: Bottomless glass bottle. Contents are inside inserted fused cone.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d5d7e4214ea827269282103a6cd8eb719acfa407&vid=0313de75c61acf9c90e590be038a7a17fba42905&prid=web&clid=SAPDAM