Imprimer
Multi-channel pipettes, mechanical, variable volume, PIPETMAN®
Fournisseur: Gilson
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PIPETMAN®
GILSF144072CEA 0 EUR
GILSF144072C GILSF144071C GILSF144070C GILSF144069CG GILSF144068CG 613-7155 GILSF144075C 613-7154 GILSF144074C 613-7153 613-7152 GILSF144073C GILSF144069C GILSF144068C GILSF144070CG 613-7159 613-7158 GILSF144072CG 613-7157 613-7156 GILSF144071CG GILSF144074CG GILSF144073CG GILSF144075CG
Multi-channel pipettes, mechanical, variable volume, PIPETMAN®
Pipettes Pipetteurs
Mechanical air-displacement pipette which offers robustness, accuracy and precision as well as a very low pipetting force for the highest comfort.
  • Very low pipetting forces thanks to easy glide piston displacement
  • Colour-coded push button ensures easy identification and GLP compliance
  • Made of PVDF and stainless steel
  • Equipped with clip ejector for convenient access to the serviceable parts
  • Blue ring positioned under the thumbwheel allows to identify PIPETMAN® and to distinguish it from the Classic and Neo models
Pipettes are individually calibrated and delivered with a certificate of conformity. Each pipette bears an engraved identification number for traceability.
Informations de commande: Models equipped with a tip ejector are only sold with a stainless steel tip ejector. Plastic tip ejectors are available in the accessories.