Imprimer
Hand sanitiser gel
Fournisseur: Deb-STOKO
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ISG100MLMD ISG1LMD ISG400MLMD
EVONISG100MLMDEA 59.8 EUR
EVONISG100MLMD EVONISG1LMD EVONISG400MLMD
Hand sanitiser gel
Soin de la peau Crèmes pour la peau Lotions pour les mains et le corps
Deb stoko sanitise gel dispenser can be used in office and facility, industrial, food, hospitality and education sectors.
  • Glove compatible - can be applied to hands prior to wearing latex, vinyl and nitrile gloves
  • No water required - quickly dries on hands without the need for rinsing with water
Deb® InstantGel advantages include highly effective formula is fully tested and proven to be bactericidal, virucidal, yeasticidal and mycobactericidal. Kills 99,999% of many common germs, rapidly and easily rubs across hands, providing the user with complete control of the product, preventing the dripping and splashing associated with using liquid alcohol sanitisers, the smooth gel is extremely pleasant for frequent use in-between hand washing, encouraging maximum compliance due to the moisturising agents glycerin and panthenol, independently tested and proven excellent skin compatibility, formulated to have low allergic potential to the skin and assessed by an independent toxicologist to be declared as 'skin hypoallergenic'.
Alcohol-based gel hand sanitisers are perfume-free and dye-free. Ideal for people who are sensitive to perfumes and dyes and who prefer products which do not have these added. When used as directed in food production and preparation areas, the product does not influence the quality of foodstuffs or present any risk to the health of its consumers.These sanitiser helps to prevent the skin from softening when sweating during glove use.
Attention: Utilisez les biocides en toute sécurité. Toujours lire l'étiquette et les informations du produit avant utilisation.