Imprimer
MitoView™, vert colorant pour mitochondries, pour microscopie de fluorescence
Marques : Biotium
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
70054 70054-T
70054.EA 210 EUR
70054. 70054-T
MitoView™, vert colorant pour mitochondries, pour microscopie de fluorescence
MitoView™
MitoView™ dyes are fluorogenic mitochondrial stains for live cells. The dyes rapidly accumulate in mitochondria and can be imaged without washing. They are available with blue, green, far-red, and near-infrared fluorescence.
  • Cellular localization: Mitochondria
  • For live or fixed cells: For live/intact cells
  • Assay type/options: No-wash staining, Real-time imaging
  • Cell permeability: Membrane permeant
  • Abs/Em: 490/523 nm
  • Detection channel: FITC, GFP
  • Potential-dependent?: No
MitoView™ Green is a green fluorescent mitochondrial dye with properties similar to those of MitoTracker® Green FM. The dye is non-fluorescent until it partitions into the mitochondrial membrane. The staining relies on mitochondrial mass, not on mitochondria membrane potential. Thus, the dye can be used to stain mitochondria in both live cells and fixed cells.* MitoView™ Green is spectrally similar to FITC.
*Note: For optimal staining of mitochondria in fixed cells or tissue sections, we recommend using one our Mitochondrial Marker Antibodies, available with a wide selection of bright and photostable CF® dyes and other conjugations.
Note: MitoView™ Green may be somewhat membrane potential-dependent in yeast cells.