Colorimetric rapid tests, sulphate, VISOCOLOR® ECO
Marques : MACHEREY-NAGELTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Determination of sulphate by turbidity measurement.
- Portable
- Environmentally friendly
- Convenient handling
- Application-oriented measuring ranges
The determination of sulphate ions is of special importance to evaluate the aggressiveness of a water towards concrete. Sulphate, a component of natural water, is formed in many manufacturing processes and is found in waste waters even after neutralisation.
Suitable for sea water testing after dilution.