Colorimetric rapid tests, nickel, VISOCOLOR® ECO
Marques : MACHEREY-NAGELTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
In ammonia solution, nickel ions react with diacetyldioxime after oxidation with bromine to form a reddish-brown dye.
- Portable
- Environmentally friendly
- Convenient handling
- Application-oriented measuring ranges
Nickel can be present in industrial waste water. It occurs as the divalent ion or as a nickel complex.
Insoluble nickel compounds (e. g. nickel cyanide, nickel carbonate) and the nickel cyano complexes are not determined.
Suitable for sea water testing after dilution.