Imprimer
Colorant selon Wright solution
Marques : Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Danger
Synonyme(s): Eosin Methylene blue solution according to Wright, Wright's eosin methylene blue
1.01383.0500 1.01383.2500 1.01383.1022 1.01383.9180 1.01383.9025
1.01383.0500EA 31.8 EUR
1.01383.0500 1.01383.2500 1.01383.1022 1.01383.9180 1.01383.9025
Colorant selon Wright solution
Colorant selon Wright
Used to stain blood, bone marrow smears, and clinical cytological specimens
The staining method according to Wright is one of the standard techniques in hematological diagnostic procedures. Like Leishman's eosin methylene blue solution and May-Grünwald's eosin methylene blue solution, it is used for staining of blood and bone marrow smears and clinical cytological specimens. The Wright’s eosin methylene blue solution is an IVD / CE product and 500 ml are sufficient for staining up to 1600 slides in Hellendahl cells.
Densité: 0.80 g/cm3 (20 °C)
Point éclair: 9.7 °C
Numéro MDL: MFCD00082143
Numéro CAS: 68988-92-1
UN: 1230
ADR: 3 (6.1) II
IVDLogo
Commander maintenant
Résultats des tests de spécification
Suitability for microscopy (Blood smear) passes test
Erythrocytes pink to brownish
Nuclei blue-violet
Eosinophilic granules red to red-brown
Neutrophilic granules light violet
Lymphocyte cytoplasm Blue