Imprimer
Handheld digital microscopes, Dino-Lite Special Light, fluorescence
Fournisseur: Dino-Lite / IDCP B.V.
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AM4115T-CFVW AM4115T-GFBW AM4115T-YFGW AM4115T-RFYW AM4115T-DFRW AM4515T4-GFBW
IDCPAM4115T-CFVWEA 852 EUR
IDCPAM4115T-CFVW IDCPAM4115T-GFBW IDCPAM4115T-YFGW 630-2749 IDCPAM4115T-DFRW IDCPAM4515T4-GFBW
Handheld digital microscopes, Dino-Lite Special Light, fluorescence
Microscopes
These fluorescence microscopes are considered to be the world’s smallest fluorescence microscopes. Compared to the traditional fluorescence microscopes with the band-pass type of emission filters, the Dino-Lites long-pass emission filters provide visibility and sensitivity over a larger range of fluorescence wavelengths.
  • Visualise fluorescence from 400 nm to 620 nm
  • High speed USB 2.0 output
  • Exchangeable caps provide more flexibility
  • Wireless streaming to a tablet, smartphone, or computer is possible in combination with WF-10 unit (to be ordered separately)
Automatic Magnification Reading (AMR): AMR displays the current magnification on screen with the included software for Windows or Mac OS. The magnification can also be automatically stored with the captured picture, making it especially useful for applications when reading the magnification is unfeasible or difficult.
Informations de livraison: Supplied with carrying pouch and software (DinoCapture 2.0 (Windows), DinoXcope (Mac OS)).