Imprimer
ESD safety shoes, Velcro® fastening, uvex 2 xenova® S1P Trainer, 9559
Fournisseur: UVEX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
9559.1/49
UVEX9559.1/49EA 0 EUR
UVEX9559.1/49
ESD safety shoes, Velcro® fastening, uvex 2 xenova® S1P Trainer, 9559
Chaussures et bottes
Metal-free, extremely lightweight, flexible and multifunctional safety shoe made from breathable water resistant high-tech micro velour upper, soft padded collar, plus a dual-density PUR sole (oil and petrol resistant, anti-static) free of paint-wetting impairment substances, silicones and plasticizers.
  • Climazone technology for improved comfort
  • Double individually adjustable velcro
  • Breathable distance mesh lining
  • Removable anti-static comfort insole with moisture control system; with a moisture-wicking system
  • Anti-slip sole with shock-absorption at heel and front of the foot
  • 100% metal-free xenova® toecap
  • Flexible, metal-free, puncture/penetration resistant midsole
  • Fulfils ESD requirements: Resistance to ground inferior to 35 megaohms
Suitable for people allergic to chrome, made from synthetic textiles.
Colour: Black/green
Certifications: EN 61340 (ESD); EN ISO 20345:2011; S1P, SRC