Imprimer
Biological indicators, self-contained tests, STERILtest
Fournisseur: LIOFILCHEM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
91100 91101 20199 91150
91100.EA 569 EUR
91100. 91101. 20199-VWR 91150.
Biological indicators, self-contained tests, STERILtest
Indicateurs de stérilisation
Biological indicators for validating the sterilisation using steam sterilisation (autoclaves).
Devices with nutrient medium, pH indicator and discs with spores of Geobacillus stearothermophilus (GST) for steam sterilisation or Bacillus atropheus (BAT) for ethylene oxide sterilisation.