Imprimer
Swabs for monitoring of surface, Contam
Fournisseur: LIOFILCHEM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
86102 86100 86103 86101 86105 86104
86102.EA 151 EUR
86102. 86100. 86103. 86101. 86105. LIOF86104
Swabs for monitoring of surface, Contam
Monitoring Systems Hygiene Monitoring Systems
For microbial monitoring of pathogens on surfaces.
  • Easy to use
  • Clear colour changes
  • No additional reagents
  • Results in 18/24 hours
  • 10 CFU/ml detection limit