Imprimer
Poly(methyl methacrylate) PMMA, analytical standard, for GPC
Marques : Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
81489-500MG 81497-500MG 81498-500MG 81501-500MG 81503-500MG 81506-1EA 81512-500MG 03558-250MG 94131-500MG 370037-25G
81489-500MGEA 299 EUR
81489-500MG 81497-500MG 81498-500MG 81501-500MG SUPL81503-500MG 81506-1EA 81512-500MG 03558-250MG 94131-500MG 370037-25G
Poly(methyl methacrylate) PMMA, analytical standard, for GPC
Étalons Étalons de chromatographie gazeuse
Poly(methyl methacrylate) is a rigid and transparent type of polymer material. It is obtained via polymerization of methyl methacrylate monomer.
Certifications: Exact values can be taken from the accompanying certificate of analysis
Informations de commande: Set (81506) contains 12 different poly(methyl methacrylate) standards with Mp ~505, ~1020, ~1960, ~5090, ~10900, ~31800, ~58700, ~103000, ~306000, ~579000, ~903000, ~2740000; always 4 standards are lyophilised in a 1.5 mL vial (green, red or white); for 1 complete calibration use one of each of the green, red and white vials
Attention: Storage temperature: 2...8 °C. Limited shelf life, expiry date on the label.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f85049f1ecc0537538355e98cc9c9da20a8e79e7&vid=f85049f1ecc0537538355e98cc9c9da20a8e79e7&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a52641ac09663148e67065d3cff2512675d2002d&vid=b17ef29ef6d36b2b1e2e1d57604c287bcd1b5002&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82e98fcb91131bab2c93a5ca97359f70e6d23f97&vid=82e98fcb91131bab2c93a5ca97359f70e6d23f97&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ede75a0e6672f73569c427d6cc2c8e37bb1d217&vid=8ede75a0e6672f73569c427d6cc2c8e37bb1d217&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d17a6299d42d74a4353984576e6d3762b28e475&vid=0d17a6299d42d74a4353984576e6d3762b28e475&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=736fea26abdb22f23eeac531cfb56434e00b4985&vid=736fea26abdb22f23eeac531cfb56434e00b4985&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=047a18f8fe0b3ed8a0c7815d1adb2b10a80477ed&vid=047a18f8fe0b3ed8a0c7815d1adb2b10a80477ed&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b7b65256ce466442521aec7aa55a0579d58b84ce&vid=b7b65256ce466442521aec7aa55a0579d58b84ce&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=53399da3f2f3b011984d23353ee041ecbae8d7e9&vid=225cb4891525966e338d7a1bf5d06f25fca759b2&prid=web&clid=SAPDAM