Imprimer
Réactif de Schiff Histology, Sigma-Aldrich®
Marques : Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Attention
1.09033.0500 1.09033.2500
1.09033.0500EA 80 EUR
1.09033.0500 1.09033.2500
Réactif de Schiff Histology, Sigma-Aldrich®
Réactif de Schiff
Ready-to-use staining solution for human-medical cell diagnosis.
Schiff‘s reagent for the periodic acid Schiff, or PAS reaction, is used for the detection of mucopolysaccharides, glycogen, muco- and glycoproteins, glyco-, phospholipids, basal membranes and collagen. With Schiff's reagent aldehydes give a bright red color reaction, thus in combination with the periodic acid the nuclei appear blue. 500 ml are sufficient for approx. 2500 - 3000 stainings. It is registered as IVD and CE certified product and can be used for clinical diagnostics.
Densité: 1.01 g/cm3 (20 °C)
Température de stockage: Température ambiante
Numéro MDL: MFCD00081985
UN: 1789
ADR: 8 II
IVDLogo