Imprimer
Test sieves, 100×40 mm
Fournisseur: Retsch GmbH
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
60.306.001000 60.306.000075 60.306.000100 60.306.000180 60.306.000280 60.306.000400
RETS60.306.001000EA 210 EUR
RETS60.306.001000 RETS60.306.000075 RETS60.306.000100 RETS60.306.000180 RETS60.306.000280 RETS60.306.000400
Test sieves, 100×40 mm
Tamis Tamis analytiques
Sieves made from 304 stainless steel are individually laser engraved with clear and precise labelling for full traceability. They are fully compatible with Retsch's existing products and can be combined with other sieves without problems.
  • Sieve bodies are made in one piece and the absence of hollow spaces prevents cross-contamination
  • High corrosion resistance and easy to clean
  • Maximum stability and optimal sealing of the sieve stack, 15% less weight than traditional sieves
Special certificates available on request. Contact your local Avantor sales office for further details.
Other mesh sizes available on request.
Certifications: ISO 3310-1:2001-09. Inspection certificate according to EN10204 3.1.B