Imprimer
ESD safety shoes, lace-up, uvex 2, xenova®, 9506
Fournisseur: UVEX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
9506.9/38 9506.9/50
UVEX9506.9/38EA 0 EUR
UVEX9506.9/38 UVEX9506.9/50
ESD safety shoes, lace-up, uvex 2, xenova®, 9506
Chaussures et bottes
Very lightweight water repellent uvex 2 xenova® shoes are made from full-grain smooth leather upper, soft padded collar and a slip-resistant Xenova® dual-density PU sole resistant to heat, chemicals, oils and gasoline. All sole materials are free of silicones, plasticisers and other paint-wetting impairment substances and suitable for people allergic to chrome.
  • Compact design, anatomical shape, good lateral stability, no thermal conductivity, non-magnetic
  • Anti-twist rear cap provides more stability and kink protection
  • Climazone technology for improved comfort
  • Anti-static comfort insole with moisture control system; orthopaedic insoles possible
  • Heat-resistant loop lacing system
  • DUO-PUR sole, non-marking, with excellent cushioning and abrasion-resistant outsole
  • Ergonomically designed outsole made from dual-density polyurethane with excellent slip resistance
  • Shock absorbing heel and forefoot
  • uvex xenova® composite toe cap (200 J)
  • Fulfils the ESD requirement: Discharge resistance <35 megohm
Used for medium-duty applications.
Colour: Black/red
Certifications: EN 61340 (ESD); EN ISO 20345:2011, S2, SRC