Imprimer
ESD safety shoes, lace-up, uvex 2 xenova®, 9554
Fournisseur: UVEX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
9554.7/38 9554.7/39 9554.7/40 9554.7/41 9554.7/42 9554.7/43 9554.7/44 9554.7/45 9554.7/46 9554.7/47 9554.7/48 9554.7/49 9554.7/50 9554.7/51 9554.7/52 9554.8/38 9554.8/39 9554.8/40 9554.8/41 9554.8/42 9554.8/43 9554.8/44 9554.8/45 9554.8/46 9554.8/47 9554.8/48 9554.8/49 9554.8/50 9554.8/51 9554.8/52 9554.9/38 9554.9/39 9554.9/40 9554.9/41 9554.9/42 9554.9/43 9554.9/44 9554.9/45 9554.9/46 9554.9/47 9554.9/48 9554.9/49 9554.9/50 9554.9/51 9554.9/52
UVEX9554.7/38EA 105 EUR
UVEX9554.7/38 UVEX9554.7/39 UVEX9554.7/40 UVEX9554.7/41 UVEX9554.7/42 UVEX9554.7/43 UVEX9554.7/44 UVEX9554.7/45 UVEX9554.7/46 UVEX9554.7/47 UVEX9554.7/48 UVEX9554.7/49 UVEX9554.7/50 UVEX9554.7/51 UVEX9554.7/52 UVEX9554.8/38 UVEX9554.8/39 UVEX9554.8/40 UVEX9554.8/41 UVEX9554.8/42 UVEX9554.8/43 UVEX9554.8/44 UVEX9554.8/45 UVEX9554.8/46 UVEX9554.8/47 UVEX9554.8/48 UVEX9554.8/49 UVEX9554.8/50 UVEX9554.8/51 UVEX9554.8/52 UVEX9554.9/38 UVEX9554.9/39 UVEX9554.9/40 UVEX9554.9/41 UVEX9554.9/42 UVEX9554.9/43 UVEX9554.9/44 UVEX9554.9/45 UVEX9554.9/46 UVEX9554.9/47 UVEX9554.9/48 UVEX9554.9/49 UVEX9554.9/50 UVEX9554.9/51 UVEX9554.9/52
ESD safety shoes, lace-up, uvex 2 xenova®, 9554
Chaussures et bottes
Very lightweight fashionable perforated uvex 2 xenova® shoes are made from synthetic materials, soft padded collar and a slip-resistant Xenova® dual-density PU sole resistant to heat, chemicals, oils and gasoline. All sole materials are free of silicones, plasticisers and other paint-wetting impairment substances and suitable for people allergic to chrome.
  • Compact design, anatomical shape, good lateral stability, no thermal conductivity
  • Climazone technology for improved comfort
  • Breathable materials as well as the perforated upper material
  • Removable anti-static comfortable insole, with moisture transport system
  • Ergonomically designed outsole made from dual-density polyurethane with excellent slip resistance
  • Shock absorption around the heel and forefoot
  • Metal-free uvex xenova® toe cap
  • Fulfils the ESD requirement: Discharge resistance <35 megohm
Used for medium-duty applications.
Colour: Blue/black
Certifications: EN ISO (ESD) 20345:2011; S1, SRC