Imprimer
Reagent reservoir chillers
Fournisseur: HEATHROW
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
120730 120731 120732 120733 120734 120735
HEAT120730EA 166 EUR
HEAT120730 HEAT120731 HEAT120732 HEAT120733 HEAT120734 HEAT120735
Reagent reservoir chillers
Refroidisseurs Refroidisseurs de paillasses
These reagent reservoir chillers are the ice-free, non-toxic way to extend the working period and preserve integrity for temperature sensitive reagents. They are made from sonic welded, engineered PP that ensures a leak free design.
  • Reagents are kept at less than 5 °C for approximately 3 hours
  • Ice-free solution eliminates the risk of contamination from messy ice baths
  • Non-toxic blend of water and bacteriostatic agent chills and freezes quickly
  • Stackable, save space when storing in the freezer or on the bench top
  • Reusable, two-year warranty
The symmetrical cooler design allows reagent reservoirs to be mounted in either direction. Engineered custom fit for Heathrow Scientific 50 or 100 ml reservoirs to ensure a maximum contact for longer uniform cooling.
Informations de commande: Assorted packs available for color coding options.
Attention: Not intended for autoclave, do not use with liquid nitrogen.