Imprimer
Phytanic acid solution ≥96%
Marques : Cayman Chemical
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Danger
Synonyme(s): 3,7,11,15-Tetramethylhexadecanoic acid
90360-5 90360-10 90360-50 90360-25
CAYM90360-5EA 148 EUR
CAYM90360-5 CAYM90360-10 CAYM90360-50 CAYM90360-25
Phytanic acid solution ≥96%
Phytanic acid
Phytanic acid is a saturated 20-carbon branched-chain fatty acid which can only be derived from dietary sources. In cases of peroxisomal disorders, elevated levels of phytanic acid are observed, and this is linked to diseases such as infantile phytanic acid storage disease and Refsum’s disease.
Formule: C₂₀H₄₀O₂
Poids moléculaire: 312.53 g/mol
Température de stockage: Congélateur
Numéro MDL: MFCD00133772
Numéro CAS: 14721-66-5
UN: 1170
ADR: 3 II
Commander maintenant
Résultats des tests de spécification
Purity ≥96%