Imprimer
Shell vials, with TITESEAL® plug style cap, KIMBLE®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SP60965D-1 SP60965D-3 SP60965D-7
SCERSP60965D-1EA 3360 EUR
SCERSP60965D-1 SCERSP60965D-3 SCERSP60965D-7
Shell vials, with TITESEAL® plug style cap, KIMBLE®
Fioles
These shell vials are made from clear KIMAX® N-51A borosilicate glass.
  • Plain tops
  • Open bottom TITESEAL® closures, unattached
Certifications: Glass conforms to USP Type I and ASTM E438, Type I, Class B requirements.