Imprimer
ESD safety ankle boots, lace-up, Matthew Mid
Fournisseur: ELTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
7677101/40 7677101/41 7677101/42 7677101/43 7677101/44 7677101/45 7677101/46 7677101/47 7677101/48 7677102/40 7677102/41 7677102/42 7677102/43 7677102/44 7677102/45 7677102/46 7677102/47 7677102/48 7677103/40 7677103/41 7677103/42 7677103/43 7677103/44 7677103/45 7677103/46 7677103/47 7677103/48
ELTN7677101/40EA 88.9 EUR
ELTN7677101/40 ELTN7677101/41 ELTN7677101/42 ELTN7677101/43 ELTN7677101/44 ELTN7677101/45 ELTN7677101/46 ELTN7677101/47 ELTN7677101/48 ELTN7677102/40 ELTN7677102/41 ELTN7677102/42 ELTN7677102/43 ELTN7677102/44 ELTN7677102/45 ELTN7677102/46 ELTN7677102/47 ELTN7677102/48 ELTN7677103/40 ELTN7677103/41 ELTN7677103/42 ELTN7677103/43 ELTN7677103/44 ELTN7677103/45 ELTN7677103/46 ELTN7677103/47 ELTN7677103/48
ESD safety ankle boots, lace-up, Matthew Mid
Chaussures et bottes
Leather-free ESD safety ankle boots made from hydrophobised microfibre and hydrophobised textile materials, reflective materials, a fully padded bellows tongue and a dual densitiy PU/PU sole ERGO-ACTIVE 2.
  • Fabric lining for natural ventilation
  • Replaceable insole; orthopaedic insoles possible
  • Semi-orthopaedic inlay sole ESD black
  • Anatomically-shaped insole and ankle support for improved stability
  • Metal-free, puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black/red
Type 1: Suitable for larger and strong feet, short toes with wide ball, heel area and steep ball angle.
Type 2: Suitable for normal feet, long toes with medium-wide ball, heel area and flat ball angle.
Type 3: Suitable for slim feet, medium-sized toes with narrow ball, heel area and medium ball angle.
Certifications: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC, form B