Imprimer
ESD safety ankle boots, lace-up, Anthony Mid
Fournisseur: Elten
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Anthony
ELTN7675103/40EA 94.9 EUR
ELTN7675103/40 ELTN7675103/42 ELTN7675101/40 ELTN7675103/41 ELTN7675101/41 ELTN7675103/44 ELTN7675101/42 ELTN7675103/43 ELTN7675103/46 ELTN7675101/43 ELTN7675103/45 ELTN7675101/44 ELTN7675101/45 ELTN7675103/48 ELTN7675103/47 ELTN7675101/46 ELTN7675101/47 ELTN7675101/48 ELTN7675102/40 ELTN7675102/41 ELTN7675102/42 ELTN7675102/43 ELTN7675102/44 ELTN7675102/45 ELTN7675102/46 ELTN7675102/47 ELTN7675102/48
ESD safety ankle boots, lace-up, Anthony Mid
Chaussures et bottes
Leather-free, sporty and lightweight ESD safety ankle boots made from microfibre, a padded tongue and a slip resistant dual-density TPU/PU sole ERGO-ACTIVE 2.
  • Fabric lining for natural ventilation
  • Semi-orthopaedic inlay sole ESD black
  • Anatomically shaped insole and ankle support for improved stability
  • Metal-free, puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in dry areas where there is a risk of penetration from pointed and sharp objects.
Colour: Black/red
Type 1: Suitable for larger and strong feet, short toes with wide ball, heel area and steep ball angle.
Type 2: Suitable for normal feet, long toes with medium-wide ball, heel area and flat ball angle.
Type 3: Suitable for slim feet, medium-sized toes with narrow ball, heel area and medium ball angle.
Certifications: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S1P, SRC, form B