Imprimer
ESD safety shoes, sandals, Velcro® fastening, Impulse EA Easy
Fournisseur: ELTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
7171501/40 7171501/41 7171501/42 7171501/43 7171501/44 7171501/45 7171501/46 7171501/47 7171501/48 7171502/40 7171502/41 7171502/42 7171502/43 7171502/44 7171502/45 7171502/46 7171502/47 7171502/48 7171503/40 7171503/41 7171503/42 7171503/43 7171503/44 7171503/45 7171503/46 7171503/47 7171503/48
ELTN7171501/40EA 91.9 EUR
ELTN7171501/40 ELTN7171501/41 ELTN7171501/42 ELTN7171501/43 ELTN7171501/44 ELTN7171501/45 ELTN7171501/46 ELTN7171501/47 ELTN7171501/48 ELTN7171502/40 ELTN7171502/41 ELTN7171502/42 ELTN7171502/43 ELTN7171502/44 ELTN7171502/45 ELTN7171502/46 ELTN7171502/47 ELTN7171502/48 ELTN7171503/40 ELTN7171503/41 ELTN7171503/42 ELTN7171503/43 ELTN7171503/44 ELTN7171503/45 ELTN7171503/46 ELTN7171503/47 ELTN7171503/48
ESD safety shoes, sandals, Velcro® fastening, Impulse EA Easy
Chaussures et bottes
Leather-free, ESD safety shoe made from microfibre, reflective materials, a padded tongue and a slip resistant dual-density TPU/PU sole ERGO-ACTIVE 2.
  • Fabric lining for natural ventilation
  • Full-length semi-orthopaedic inlay sole ESD black
  • Soft fleece ESD insole
  • Anatomically shaped insole and ankle support for improved stability
  • Protective steel toecap (200 J)
Ideal for use in dry work areas such as industry, storage, transport, construction etc.
Colour: Black/blue
Type 1: Suitable for larger and strong feet, short toes with wide ball, heel area and steep ball angle.
Type 2: Suitable for normal feet, long toes with medium-wide ball, heel area and flat ball angle.
Type 3: Suitable for slim feet, medium-sized toes with narrow ball, heel area and medium ball angle.
Certifications: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S1, SRC, form A