Imprimer
Cleanroom documentation papers and notebooks, CONTEXT™
Fournisseur: CONTEC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
CNBK5585W CNBK8511W CNTXA4W CNTXA4B
818-0296EA 501 EUR
818-0296 818-0297 818-0299 818-0298
Cleanroom documentation papers and notebooks, CONTEXT™
Papiers
CONTEXT™ cleanroom documentation products have been developed to provide cleanrooms with paper and notebooks that reduce the risk of contamination.
  • Compatible with standard and high speed laser printers
  • Compatible with wet and dry copy systems
  • Ideal for recording notes and work instructions
  • High brightness, high opacity
  • Ultra-low particle generation
  • Ultra-low metallic ion content
  • High tear and tensile strength
  • Heat resistant
CONTEXT™ cleanroom paper is available in A4, 216×279 mm and 279×432 mm sizes.
CONTEXT™ notebooks are available in two sizes. They are spiral bound and protected with durable plastic covers. Pages are college ruled, with 50 sheets per notebook.
It uses Munising® Bond material made from synthetic polymer-saturated cellulose to provide optimal smoothness and natural enhanced writability. Double-bagged to ensure product integrity and uncompromised entry into a sterile suite.