Imprimer
ESD safety shoes, lace-up, impulse EA blue low, S1
Fournisseur: ELTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
7271501/47 7271501/40 7271501/41 7271501/42 7271501/43 7271501/44 7271501/45 7271501/46 7271501/48 7271502/40 7271502/41 7271502/42 7271502/43 7271502/44 7271502/45 7271502/46 7271502/47 7271502/48 7271503/40 7271503/41 7271503/42 7271503/43 7271503/44 7271503/45 7271503/46 7271503/47 7271503/48
ELTN7271501/47EA 86.9 EUR
ELTN7271501/47 ELTN7271501/40 ELTN7271501/41 ELTN7271501/42 ELTN7271501/43 ELTN7271501/44 ELTN7271501/45 ELTN7271501/46 ELTN7271501/48 ELTN7271502/40 ELTN7271502/41 ELTN7271502/42 ELTN7271502/43 ELTN7271502/44 ELTN7271502/45 ELTN7271502/46 ELTN7271502/47 ELTN7271502/48 ELTN7271503/40 ELTN7271503/41 ELTN7271503/42 ELTN7271503/43 ELTN7271503/44 ELTN7271503/45 ELTN7271503/46 ELTN7271503/47 ELTN7271503/48
ESD safety shoes, lace-up, impulse EA blue low, S1
Chaussures et bottes
Leather-free, sporty and lightweight ESD safety shoes made from microfibre, reflective materials, a full padded bellows tongue and a slip resistant dual-density TPU/PU sole ERGO-ACTIVE 2.
  • Fabric lining for natural ventilation
  • Replaceable insole; orthopaedic insoles possible
  • Full-length semi-orthopaedic inlay sole ESD black
  • Soft fleece ESD insole
  • Elastic lacing system with individual quick locking system
  • Protective steel toecap (200 J)
Ideal for dry work areas such as industry, storage, transport, assembly and areas where there is a risk of electrostatic discharge.
Colour: Blue/black
Type 1: Suitable for larger and strong feet, short toes with wide ball, heel area and steep ball angle.
Type 2: Suitable for normal feet, long toes with medium-wide ball, heel area and flat ball angle.
Type 3: Suitable for slim feet, medium-sized toes with narrow ball, heel area and medium ball angle.
Certifications: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S1, SRC, form A