Imprimer
ESD safety shoes, lace-up, Mason Pro Low
Fournisseur: ELTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
7281101/45 7281101/46 7281101/40 7281101/41 7281101/42 7281101/43 7281101/44 7281103/40 7281103/41 7281103/42 7281103/43 7281103/44 7281103/45 7281103/46 7281103/47 7281103/48 7281101/47 7281101/48 7281102/40 7281102/41 7281102/42 7281102/43 7281102/44 7281102/45 7281102/46 7281102/47 7281102/48
ELTN7281101/45EA 74.9 EUR
ELTN7281101/45 ELTN7281101/46 ELTN7281101/40 ELTN7281101/41 ELTN7281101/42 ELTN7281101/43 ELTN7281101/44 ELTN7281103/40 ELTN7281103/41 ELTN7281103/42 ELTN7281103/43 ELTN7281103/44 ELTN7281103/45 ELTN7281103/46 ELTN7281103/47 ELTN7281103/48 ELTN7281101/47 ELTN7281101/48 ELTN7281102/40 ELTN7281102/41 ELTN7281102/42 ELTN7281102/43 ELTN7281102/44 ELTN7281102/45 ELTN7281102/46 ELTN7281102/47 ELTN7281102/48
ESD safety shoes, lace-up, Mason Pro Low
Chaussures et bottes
Lightweight and comfortable ESD safety shoes made from cowhide leather, GORE-TEX® lining, reflective materials, a full padded bellows tongue and a dual density PU-PU sole ERGO-ACTIVE 2.
  • Fabric lining for natural ventilation
  • Semi-orthopaedic inlay sole ESD black
  • Soft fleece ESD insole
  • Anatomically-shaped insole and ankle support for improved stability
  • TPU scuff cap
  • Puncture resistant midsole made from steel
  • Protective steel toe cap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black/grey
Type 1: Suitable for larger and strong feet, short toes with wide ball, heel area and steep ball angle.
Type 2: Suitable for normal feet, long toes with medium-wide ball, heel area and flat ball angle.
Type 3: Suitable for slim feet, medium-sized toes with narrow ball, heel area and medium ball angle.
Certifications: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC, form A