Imprimer
ESD safety shoes, sandals, Velcro® fastening, Anthony Easy
Fournisseur: ELTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
7175101/40 7175101/41 7175101/42 7175101/44 7175101/45 7175101/46 7175101/47 7175101/48 7175103/40 7175103/41 7175103/42 7175103/43 7175103/44 7175103/45 7175103/46 7175103/47 7175103/48 1717511/36 1717511/37 1717511/38 1717511/39 1717513/36 1717513/37 1717513/38 1717513/39
ELTN7175101/40EA 0 EUR
ELTN7175101/40 ELTN7175101/41 ELTN7175101/42 ELTN7175101/44 ELTN7175101/45 ELTN7175101/46 ELTN7175101/47 ELTN7175101/48 ELTN7175103/40 ELTN7175103/41 ELTN7175103/42 ELTN7175103/43 ELTN7175103/44 ELTN7175103/45 ELTN7175103/46 ELTN7175103/47 ELTN7175103/48 ELTN1717511/36 ELTN1717511/37 ELTN1717511/38 ELTN1717511/39 ELTN1717513/36 ELTN1717513/37 ELTN1717513/38 ELTN1717513/39
ESD safety shoes, sandals, Velcro® fastening, Anthony Easy
Chaussures et bottes
Leather-free, sporty and lightweight ESD safety sandal made from microfibre, a padded tongue and a slip resistant dual-density TPU/PU sole ERGO-ACTIVE 2.
  • Fabric lining for natural ventilation
  • Full-length semi-orthopaedic inlay sole ESD black
  • Anatomically shaped insole and ankle support for improved stability
  • Metal-free, puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in dry areas where there is a risk of penetration from pointed and sharp objects. The sizes 36 - 39 are ideally tailored to the ergonomics of female feet.
Colour: Black/red
Type 1: Suitable for larger and strong feet, short toes with wide ball, heel area and steep ball angle.
Type 2: Suitable for normal feet, long toes with medium-wide ball, heel area and flat ball angle.
Type 3: Suitable for slim feet, medium-sized toes with narrow ball, heel area and medium ball angle.
Certifications: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S1P, SRC, form A