Imprimer
ESD safety shoes, lace-up, Mason Pro GTX Low
Fournisseur: ELTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
7283101/40 7283101/41 7283101/42 7283101/43 7283101/44 7283101/45 7283101/46 7283101/47 7283101/48 7283102/40 7283102/41 7283102/42 7283102/43 7283102/44 7283102/45 7283102/46 7283102/47 7283102/48 7283103/40 7283103/41 7283103/42 7283103/43 7283103/44 7283103/45 7283103/46 7283103/47 7283103/48
ELTN7283101/40EA 113.9 EUR
ELTN7283101/40 ELTN7283101/41 ELTN7283101/42 ELTN7283101/43 ELTN7283101/44 ELTN7283101/45 ELTN7283101/46 ELTN7283101/47 ELTN7283101/48 ELTN7283102/40 ELTN7283102/41 ELTN7283102/42 ELTN7283102/43 ELTN7283102/44 ELTN7283102/45 ELTN7283102/46 ELTN7283102/47 ELTN7283102/48 ELTN7283103/40 ELTN7283103/41 ELTN7283103/42 ELTN7283103/43 ELTN7283103/44 ELTN7283103/45 ELTN7283103/46 ELTN7283103/47 ELTN7283103/48
ESD safety shoes, lace-up, Mason Pro GTX Low
Chaussures et bottes
Lightweight and comfortable ESD safety shoes made from cowhide leather, GORE-TEX® lining, reflective materials, a full padded bellows tongue and a dual density PU-PU sole ERGO-ACTIVE 2.
  • GORE-TEX® climate-regulating, breathable textile lining, water resistance
  • Semi-orthopaedic inlay sole ESD black
  • Anatomically-shaped insole and ankle support for improved stability
  • TPU scuff cap
  • Metal-free, puncture resistant midsole
  • Protective steel toe cap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black/Blue
Type 1: Suitable for larger and strong feet, short toes with wide ball, heel area and steep ball angle.
Type 2: Suitable for normal feet, long toes with medium-wide ball, heel area and flat ball angle.
Type 3: Suitable for slim feet, medium-sized toes with narrow ball, heel area and medium ball angle.
Certifications: EN 61340-5-1(ESD); DGUV 112-191 (BGR 191); EN ISO 20345; S3 SRC, form B