Imprimer
VWR®, Chaussettes de salle blanche, non ESD, jetables
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
113-9407EA 540 EUR
113-9407 113-9408
VWR®, Chaussettes de salle blanche, non ESD, jetables
Disposable Garments
These disposable socks are made of polyester filament yarn.
  • Ultra-clean treatment
  • Low particle generation
  • Friction resistance
  • Soft and comfortable to wear
Suitable for use in ISO Class 4 to 6 (Class 10 to 1000) cleanrooms.
Used disposable socks can be recycled using the Fabrics and Clothing Zero Waste Box™ from TerraCycle.