Imprimer
ESD safety shoes, lace-up, Sensation XX10 Low
Fournisseur: ELTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
729901/36 729901/37 729901/38 729901/39 729901/40 729901/41 729901/42 729901/43 729901/44 729901/45 729901/46 729901/47 729901/48
114-9072EA 91.9 EUR
114-9072 114-9073 114-9074 114-9075 114-9076 114-9077 114-9078 114-9079 114-9080 114-9081 114-9082 114-9083 114-9084
ESD safety shoes, lace-up, Sensation XX10 Low
Chaussures et bottes
Excellent wearing comfort, with a padded upper edge to protect the Achilles tendon.
  • Hydrophobised textile material with jeans look/Hydrophobized nubuck leather
  • Fabric lining bioactive
  • Padded tongue
  • Full-length inlay sole ESD PRO grey
  • Non-metallic, penetration-resistant midsole
  • PU/PU sole WELLMAXX L10
  • Steel toe cap
Thanks to its excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA). Sole core made of Infinergy® by BASF. Extra laces included.
Colour: Black/grey
Certifications: EN 61340-5-1 (ESD), EN ISO 20345; S3, SRC, form A