Imprimer
ViVi-CR Battery for Cleanroom Hood System
Fournisseur: THI Total Healthcare Innovation GmbH
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
60402 60501
THIG60402EA 188 EUR
THIG60402 THIG60501
ViVi-CR Battery for Cleanroom Hood System
Piles
Black battery system with 49.87 Wh capacity. It charges faster, lasts longer, is equipped with a led display of charging state and ending battery acoustic alarm.
White small and lightweight Li-Ion battery pack with 43.6 Wh capacity. The charger holds four battery units.