Imprimer
Muffle furnaces with lift up door, Tmax 1300 °C and standard controller B510, LT 5/13 - LT 15/13
Fournisseur: NABERTHERM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
L-055H3ANL L-055H3ANLG L-055H3ANLGT L-055H3ANLGTZ L-055H3ANLGZ L-055H3ANLT L-055H3ANLTZ L-055H3ANLZ
NABEL-055H3ANLEA 5350 EUR
NABEL-055H3ANL NABEL-055H3ANLG NABEL-055H3ANLGT NABEL-055H3ANLGTZ NABEL-055H3ANLGZ NABEL-055H3ANLT NABEL-055H3ANLTZ NABEL-055H3ANLZ
Muffle furnaces with lift up door, Tmax 1300 °C and standard controller B510, LT 5/13 - LT 15/13
Étuves Fours
These furnaces feature heating elements on supporting tubes that radiate freely into the chamber, ensuring particularly short heating times. With the robust lightweight refractory brick insulation, they can reach a max. working temperature of 1300 °C.
  • Heating from two sides
  • Multi-layer insulation with robust lightweight refractory bricks in the furnace chamber
  • Textured stainless steel housing
  • Dual shell housing for low external temperatures and high stability
  • Lift up door (LT) with hot surface facing away from the operator
  • Exhaust air outlet on rear wall of furnace
The furnace is equipped with the B510 standard controller with touch screen display, featuring 5 freely adjustable programs and 4 segments and 2 extra functions. These controllers are PID microprocessor controlled with a self-diagnostic system and integrated process documentation. The B510 controllers can automatically activate the vent with fan or catalyst (not model LT 15).
Informations de commande: All units require a 230 V single-phase power supply. Optional accessories e.g. a vent, vent with fan or catalytic converter, over-temperature limit controller with adjustable temperature cut out for thermal protection Class 2, in accordance with EN 60519-2 as temperature limiter to protect the oven and load, protective gas connection on the rear wall of the furnace. Manual or automatic gas supply systems are also available. Corresponding models with fold down doors, different Tmax and other controllers are also available. Please search the website for more details.
Attention: Maximum continuous operating temperature should be 50 °C below the 'Max. temp.' of the model.
Outer dimensions include opened lift up door.
*Temperature uniformity: ± 5 K with closed air inlet in the empty workspace according to DIN 17052-1 at working temperatures > 800 °C