Imprimer
Muffle furnaces with lift up door, Tmax 1200 °C and standard controller B510, LT 3/12 - LT 60/12
Fournisseur: NABERTHERM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
L-154H2LN2TZ
455-1490EA 0 EUR
455-1490
Muffle furnaces with lift up door, Tmax 1200 °C and standard controller B510, LT 3/12 - LT 60/12
Étuves Fours
The LT 3/12 - LT 60/12 muffle furnaces are the ideal choice for daily laboratory use. These high quality models have an advanced and attractive design, and are very reliable. The muffle furnaces have a lift up door as standard.
  • Textured stainless steel housing
  • Dual shell housing for low external temperatures and high stability
  • Lift up door (LT) with hot surface facing away from the operator
  • Exhaust air outlet on the rear wall of furnace
  • Solid state relays provide low-noise operation
  • Defined application within the constraints of the operating instructions
  • NTLog Basic for Nabertherm controller: Recording of process data with USB-flash drive
Heating is from two sides by means of ceramic heating plates (heating from three sides for muffle furnaces LT 24/12 - LT 60/12). These ceramic heating plates with an integral heating element are protected from fumes and splashing, and are easy to replace. Only fibre materials which are not classified as carcinogenic according to TRGS 905, class 1 or 2 are used.
The furnace is equipped with a B510 standard controller with touch screen display, featuring 5 freely adjustable programs and 4 segments and 2 extra functions. These controllers are PID microprocessor controlled with a self-diagnostic system and integrated process documentation. The B510 controllers can automatically activate the vent with fan or catalyst (not model LT 15).
Informations de commande: All units require a 230 V single-phase power supply, except LT 24, LT 40 and LT 60 (400 V 3/N/PE). Optional accessories e.g. a vent, vent with fan or catalytic converter, over-temperature limit controller with adjustable temperature cut out for thermal protection Class 2, in accordance with EN 60519-2 as temperature limiter to protect the oven and load, protective gas connection on the rear wall of the furnace. Manual or automatic gas supply systems are also available. Corresponding models with fold down doors, different Tmax and other controllers are also available. Please search the website for more details.
Attention: Maximum continuous operating temperature should be 50 °C below the 'Max. temp.' of the model.
Note heat up times: If connected at 230 V 1/N/PE rsp. 400 V 3/N/PE.
Outer dimensions include opened lift up door.
* Temperature uniformity of ± 5K in the empty work space for temperatures >800 °C.