Imprimer
Adapters, PTFE, GL thread, BOLA
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
D730-24 D730-12
BOHLD730-24EA 96.1 EUR
BOHLD730-24 BOHLD730-12
Adapters, PTFE, GL thread, BOLA
Adaptateurs Raccords à rodage conique
Adaptor made of glass-fibre reinforced PTFE.
  • Transition from pump thread M20x1,5 to GL thread
  • Pressure resistant connection of maximum 10 bar of hardwalled tubing
  • Universal chemical resistance
  • Temperature resistant from –200 to +250 °C