Imprimer
BOD calibration standards, (CRM), Supelco®
Marques : Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BOD1000-500ML BOD2000-500ML BOD200-100ML BOD200-500ML BOD500-500ML
BOD1000-500MLEA 128 EUR
BOD1000-500ML BOD2000-500ML BOD200-100ML BOD200-500ML BOD500-500ML
BOD calibration standards, (CRM), Supelco®
Étalons Étalons pour l'analyse environnementale
Certifications: These certified reference materials (CRM) are produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.
Attention: Storage temperature: 2 - 8 °C (refrigerator)