Imprimer
BCECF AM ester ≥95% (par HPLC) indicateur de pH fluorescent
Marques : Biotium
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Synonyme(s): BCECF AM
51011 51011-1 51012
51011.EA 217 EUR
51011. 51011-1 51012.
BCECF AM ester ≥95% (par HPLC) indicateur de pH fluorescent
BCECF AM ester
BCECF, AM ester is membrane-permeant and thus can be loaded into cells via incubation. This product is a mixture of three molecular species, all of which are readily hydrolyzed by endogenous esterases into a single BCECF free acid form once they are in the cells.
  • Cell permeability: Membrane permeant
  • Colors: Green
  • Absorption/Emission 508 nm/530 nm (for BCECF acid)
  • White solid soluble in DMSO and DMF
  • Store at -20°C and protect from light, especially in solution
Formule: C₄₂H₄₀O₂₁
Poids moléculaire: 808.69 g/mol
Température de stockage: Congélateur
Numéro MDL: MFCD00036969
Numéro CAS: 117464-70-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=825a7550c456e5d203606af9dc6266ba39c0ff67&vid=9a4aca4f3bc9c03b7d450afef197f168ffbde83c&prid=web&clid=SAPDAM