Imprimer
Waste bins, WIVA™ VAT-5
Fournisseur: HANNELLS INDUSTRIER
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
030V5090BL 050V5090BL
HANL030V5090BLEA 0 EUR
HANL030V5090BL HANL050V5090BL
Waste bins, WIVA™ VAT-5
Poubelles Conteneurs de déchets
WIVA™ VAT-5 medical waste containers provide safe storage and transport of healthcare hazardous waste.
  • Fit for use in various medical environments, also for hygienic autoclaving and incineration processes
  • Simple but robust closing system
Certifications: UN approved for Class 6.2 infectious waste, UN 3291