Imprimer
Empty FPLC columns, G-TRAP™
Fournisseur: G-Biosciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
786-1291 786-1290
GENO786-1291EA 58.5 EUR
GENO786-1291 GENO786-1290
Empty FPLC columns, G-TRAP™
Colonnes pour chromatographie
These empty columns are designed for fast protein liquid chromatography.
  • Frit material is ultra-high molecular weight polyethylene
Columns are made of polypropylene and come in 1 ml and 5 ml sizes.
Informations de livraison: Includes column tube, sealing plug, sealing sleeve, top stop plug and bottom stop plug, top (large) frit, bottom (small) frit (inserted in column).